Learn More
Thermo Scientific™ EPD Mk2, G and N2 Readers
Product Code. p-4088856 Shop All Thermo Scientific Products
Click to view available options
Weight (Metric):
50 g
Length (Metric) Cable:
1 m
Unit Size:
Each
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11927927
Brand: Thermo Scientific™ EPD162570500
This item requires a price update prior to being ordered.
Request a Price for this item here.
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
To complement the Thermo Scientific EP DMk2, G and N2. The basic IrDA reader, is ideal for office environments and mobile applications
Compatible with the full range of Mk2 software products
  • Connect directly to the PC via a serial or optional USB interface
  • Require no power supply
  • For more demanding environments, and where it is necessary to ensure that only one specific EPD can be “seen” by the reader, the desktop model ACT-5 may be used
  • The IrDA and ACT-5 are compatible with EasyEPD2 and EasyIssue software
Specifications
Weight (English) 0.1 lb.
Weight (Metric) 50 g
Connector Types Serial RS232 or USB 1.1 and 2.0 compatible interface direct to PC
For Use With (Equipment) EasyEPD2 and EasyIssue software
For Use With (Application) For office Environments and Mobile applications
Includes Easy EPD software, USB interface
Length (English) Cable 3.2 ft.
Length (Metric) Cable 1 m
Product Type Basic Irda Reader