Description
Increase efficiency, enable portability
Specifications
Specifications
| For Use With (Equipment) | Steri-Cult CO2 3310 incubator |
| Type | Shelf Rack |
| Width (English) | 8.6 in. |
| Width (Metric) | 21.8 cm |
| Height (English) | 6.6 in. |
| Height (Metric) | 16.8 cm |
| Depth (English) | 19.8 in. |
| Depth (Metric) | 50.29 cm |