Learn More
Thermo Scientific™ H-6000A Bucket Adapters
Accept a variety of blood processing tubes, bags and bottles
Brand:  Thermo Scientific™ 00444
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 13144299
  • 423.00€ / Each
Estimated Shipment: 27-08-2024
to see stock.
Description
Description
Accept a variety of blood processing tubes, bags and bottles
For use with Thermo Scientific H-6000A buckets.
  • Adapters hold a maximum capacity of 6 × 500mL (bottles), 6 × 550mL (blood bags), and 132 × 7mL (tubes). Max. O.D. × L: 70 x 160mm
Specifications
Specifications
6 x 500 mL Bottles
H-6000A bucket for Thermo Scientific™ Sorvall™ RC3BP™ Plus low speed centrifuge rotor