missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Hypersil™ Prism RP Columns--5μm Particle Size

Offered in lengths from 50 to 250mm, with I.D. from 1.0 to 20mm

599.76€ - 645.66€

Specifications

Particle Size 5 μm
USP Type L60
Stationary Phase Alkyl Amide
View More Specs

Products 2
Product Code Brand Diameter (Metric) Length (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Length (Metric) Price Quantity & Availability  
11512921
View Documents Promotion Details
Thermo Scientific™
32105-102130
2.1 mm 100 mm This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
11502921
View Documents Promotion Details
Thermo Scientific™
32105-052130
2.1 mm 50 mm This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

5 μm
L60
Alkyl Amide
Videos
SDS
Documents

Documents

Product Certifications