Learn More
Thermo Scientific™ Lab Vision™ Dewax and HIER Buffer L,M,H (IVD)
Thermo Scientific Dewax and Heat Induced Epitope Retrieval (HIER) Buffers simultaneously automate deparaffinization, rehydration and epitope retrieval in the Thermo Scientific PT Module.
Brand:  Thermo Scientific™ TA-100-DHBM
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15270705
  • 2175.00€ / 100mL
Estimated Shipment date
from Supplier 22-06-2020
Add to basket
Description
Description
Automated deparaffinization (dewaxing) and rehydration of IHC slides is an ideal way to decrease turn-around time and improve lean workflow. Aqueous deparaffinization is the method of choice for IHC instruments today but often yields incomplete deparaffinization. The new Thermo Scientific™ Dewax and Heat Induced Epitope Retrieval (HIER) Buffers simultaneously automate deparaffinization, rehydration and epitope retrieval in the Thermo Scientific PT Module. The new proprietary formulation eliminates manual steps and improves turnaround time. In addition the results are concordant to traditional methods meaning they can be quickly integrated into routine workflow.
Specifications
Specifications
Buffer Medium
100mL