Learn More
Thermo Scientific™ Owl™ Accessories for Giraffe™ Adjustable Gel System
Product Code. 12454687
Click to view available options
Thickness (Metric):
0.08 cm
0.32 cm
Width (Metric):
0.1 cm
16 cm
Type:
Blank Glass Plate
Notched Glass Plate
Spacer Set
Unit Size:
Each
Product Code. 12454687
Brand: Thermo Scientific™ P516G
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Glass and spacer set
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) ADJ1 Giraffe Adjustable Gel Electrophoresis System
Type Blank Glass Plate
Material Glass
Thickness (English) 0.12 in.
Thickness (Metric) 0.32 cm
Length (Metric) 16 cm
Width (Metric) 16 cm