Learn More
Thermo Scientific™ QCC (Quick Column Change) for Agilent Instruments
Product Code. 13446956 Shop All Thermo Scientific Products
Click to view available options
Description:
S/SL Injector Adaptor
TCD Detector Adaptor
Tight Nut
Unit Size:
Each
Pack of 2
Product Code. 13446956
Brand: Thermo Scientific™ 60180873
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Quick Column Change accessory is compatible with Agilent Instrumentation
QCC (Quick Column Change)-Leak Free Column Installation
  • Tool-free installation of analytical GC column
  • Graphite ferrules
Complete QCC Kits Includes:
  • Injector Adaptor
  • Detector Adaptor
  • Tight Nut
  • Ferrules (1 each 0.25, 0.32,0.53)
  • Ceramic Column Cutter
TRUSTED_SUSTAINABILITY
Specifications
Description Tight Nut
For Use With Agilent and Thermo Scientific Instruments