Learn More
Epredia™ Richard-Allan Scientific™ Cyto-Stain™ G
Shop All Epredia Products
Click to view available options
Quantity:
1 gal.
Unit Size:
Each
Description
- Can be used for all gynecological and non-gynecological procedures
- Designed to produce greener cyanophilic hues in intermediate and basal cells
- Requires one counterstain instead of two
- Reduces staining time
Specifications
Specifications
| Type | Cytostain |
| Quantity | 1 gal. |