Learn More
Thermo Scientific™ Sorvall™ Microplate Carrier
Accommodates 4 x 4 standard microplates (or one deepwell plate)
Brand:  Thermo Scientific™ 75006449
Additional Details : Weight : 1.18800kg
Product Code. 10639142
  • 590.00€ / Each
Estimated Shipment: 26-06-2024
to see stock.
Description
Description
  • For use with Four-Place Swinging Bucket Rotors 75-006-441 and 75-006-446
  • Maximum speed/RCF: 4150rpm/3060xG
  • Maximum carrier height: 60mm
Order Info
Sold individually; order four to fully equip rotor.
Specifications
Specifications
Thermo Scientific™ Sorvall™ swing bucket rotor