Learn More
Trajan Scientific and Medical™ Thermo Scientific Splitless FocusLiner (For Use with 70mm Needle)
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Brand:  Trajan Scientific and Medical™ 092045
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 12933981
  • 255.00€ / Pack of 5
Estimated Shipment: 05-07-2024
to see stock.
Add to basket
Description
Description
  • Each batch of liners tested for activity
  • Manufactured from borosilicate glass
  • Color coded by geometry for easy identification
  • For use with Thermo Scientific instruments
Specifications
Specifications
70 mm Needle
5/Pk.
5 mm
Thermo Scientific Trace and Focus GCs
GC Inlet Liner
105 mm
8 mm