Learn More
Thermo Scientific™ Versette ClipTips™
Unique interface locks tip onto Versette automated liquid handling system.
Brand:  Thermo Scientific™ 5808
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11664912
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
Tips have a clasp or clip that locks in place to ensure an effective seal and will only release when mechanically ejected.
  • Unique design – On or Off only – tips lock in place providing a secure fit and preventing loose, uneven or leaky tips
  • Minimizing cross contamination between samples, the tips reach the bottom of tubes
  • Versatile tip range with three sizes for volumes: 1250μL, 300/125μL and 30/12.5μL
  • Rigid rack allows stable, even tip seating
Specifications
Specifications
100 to 1250 μL
Non-sterile
On or off tips lock in place providing a secure fit and preventing loose, uneven or leaky tips, minimizing cross contamination between samples, provide stable, straight mounting every time
Automation
Versette ClipTips
Filtered
1250μL
Rack
Thermo Scientific Versette