Learn More
Eppendorf™ Thermomixer™ R Accessory, Thermoblock Dry Blocks
Product Code. p-3803175 Shop All Eppendorf Products
Click to view available options
Holds:
8 x 5.0 mL
For Use With (Equipment):
Eppendorf Tube
Unit Size:
Each
Product Code. 15108374
Brand: Eppendorf™ 5309000333
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Ensure precise fit for common lab tubes
These exchangeable thermoblocks for the Thermomixer R and ThermoStat provide excellent flexibility. The range of potential applications is significantly expanded due to the support for slides and microarrays, in addition to a broad range of tube sizes.
  • Shielded thermoblocks are contact-safe
  • Efficient exchange simplifies operation
  • Tubes are visible during operation and easily accessible
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Eppendorf Tube
Holds 8 x 5.0 mL
Type Block
For Use With (Application) For 8 x Eppendorf Tube 5.0 mL
Includes Not Applicable