Learn More
Sonoco™ ThermoSafe PolarPack™ Standard
Click to view available options
Dimensions (LxWxH):
10.5 x 7.5 x 1.25 in.
5.5 x 3.5 x 0.75 in.
6 x 5.5 x 1 in.
6.5 x 5.5 x 1 in.
8 x 5.5 x 1.25 in.
Weight (English):
6 oz.
12 oz.
16 oz.
20 oz.
48 oz.
Material:
Polyethylene
Polyethylene Film
Unit Size:
Pack of 12
Pack of 24
Pack of 36
Pack of 48
Pack of 768
Pack of 96
Description
- Thermal solutions for temperature critical packages during transit
- Gel will phase change at 0° C.
- condition coolant in freezer before shipments.
- refrigeration for transport of food, pharmaceuticals and medical products.
- For frozen or refrigerated shipments.
- Alternative to wet ice.
- Available in several sizes
- Cold shipping supplies .
Specifications
Specifications
| Length (English) Exterior | 5.5 in. |
| Width (English) Exterior | 3.5 in. |
| Height (English) Exterior | 0.75 in. |
| Weight (English) | 6 oz. |
| Dimensions (LxWxH) | 5.5 x 3.5 x 0.75 in. |
| Material | Polyethylene Film |
| Freezing Point | 0°C |
| For Use With (Application) | For shipping temperature critical packages during transit |