Learn More
Sonoco™ ThermoSafe PolarPack™ Standard
Product Code. 16383356
Click to view available options
Dimensions (LxWxH):
10.5 x 7.5 x 1.25 in.
5.5 x 3.5 x 0.75 in.
6 x 5.5 x 1 in.
6.5 x 5.5 x 1 in.
8 x 5.5 x 1.25 in.
Weight (English):
6 oz.
12 oz.
16 oz.
20 oz.
48 oz.
Material:
Polyethylene
Polyethylene Film
Unit Size:
Pack of 12
Pack of 24
Pack of 36
Pack of 48
Pack of 768
Pack of 96
Product Code. 16383356
Brand: Sonoco™ PP20IN24BL
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
PolarPack refrigerants provide thermal solutions for temperature critical packages during transit. Available in a broad selection of styles and a multitude of sizes, our refrigerants are designed to provide reliable temperature protection for a vast array of temperature-sensitive products and packages.
  • Thermal solutions for temperature critical packages during transit
  • Gel will phase change at 0° C
  • Condition coolant in freezer before shipments
  • Refrigeration for transport of food, pharmaceuticals and medical products
  • For frozen or refrigerated shipments
  • Alternative to wet ice
  • Quantity: Pack of 768 pieces
TRUSTED_SUSTAINABILITY
Specifications
Quantity 768/Pk.
Weight (English) 20 oz.
Dimensions (LxWxH) 8 x 5.5 x 1.25 in.
Material Polyethylene
Freezing Point 0°C
For Use With (Application) For shipping temperature critical packages during transit