Learn More
Thiamine. HCl, Serva Electrophoresis™
Product Code. 11727826
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
Unit Size:
Each
Product Code. Quantity unitSize
11727826 100 g Each
1 options
Product Code. 11727826
Brand: Serva Electrophoresis 3602002
Specifications
Color White
Density 0.4g/cm3
pH 3 (20°C)
Quantity 100 g
Solubility Information Solubility in/miscibility with Water at 20°C
Physical Form Powder