missing translation for 'onlineSavingsMsg'
Learn More

ANSELL™ Thick Natural Rubber Latex AlphaTec™ Heavy Duty Gloves

Product Code. 17963542 Shop All Ansell Products
Click to view available options
Model:
87-950
87-955
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 17963542

Brand: ANSELL™ 87950075

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Extra heavy duty black glove with 0.73 mm thickness and 100% cotton flock lining.

  • 50% thicker than standard flock lined gloves
  • Extremely high resistance to many ketones, salts, detergents, alcohols, alkalies and fats
  • Excellent resistance to mechanical hazards plus increased chemical protection
  • Chlorinated
  • 100% cotton flock lining to help absorb perspiration
  • AQL 1.5
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) CE 0493|EN 388:2016 +A1:2018|EN ISO 21420:2020|EN ISO 374-1:2016|EN ISO 374-5:2016|Category III|REACH Compliant
Chlorinated Yes
Applications Industrial
Color Black
Latex Free No
Size 7.5
EN388 Standard X121X
EN 421 No
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES AKLOPS
EN ISO 374 - 5 Bacteria, Fungi WITH Virus Protection
Glove Cuff Style Pinked
Grip Design Lozenge
Lining Unlined
Model 87-950
Length (English) 12.8 in.
Length (Metric) 325 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.