missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANSELL™ Thick Natural Rubber Latex AlphaTec™ Heavy Duty Gloves
Shop All Ansell Products
Click to view available options
Model:
87-950
87-955
Unit Size:
12 pairs
Description
- 50% thicker than standard flock lined gloves
- Extremely high resistance to many ketones, salts, detergents, alcohols, alkalies and fats
- Excellent resistance to mechanical hazards plus increased chemical protection
- Chlorinated
- 100% cotton flock lining to help absorb perspiration
- AQL 1.5
Specifications
Specifications
| Certifications/Compliance (Europe) | CE 0493|EN 388:2016 +A1:2018|EN ISO 21420:2020|EN ISO 374-1:2016|EN ISO 374-5:2016|Category III|REACH Compliant |
| Chlorinated | Yes |
| Applications | Industrial |
| Color | Black |
| Latex Free | No |
| Size | 7.5 |
| EN388 Standard | X121X |
| EN 421 | No |
| EN ISO 374-1 | Type A |
| EN ISO 374-1 CHEMICAL LETTER CODES | AKLOPS |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction