Learn More
Thiobenzoic acid, 90%, Thermo Scientific Chemicals
Catalog Number p-3758808
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
98-91-9
MFCD00004852
UIJGNTRUPZPVNG-UHFFFAOYSA-N
thiobenzoic acid, benzenecarbothioic acid, benzoyl thiol, monothiobenzoic acid, benzoic acid, thio, acido mercaptobenzoico, unii-gbg5rlo56n, ccris 8913, acido mercaptobenzoico italian, gbg5rlo56n
7414
benzenecarbothioic S-acid
C1=CC=C(C=C1)C(=O)S
98-91-9
MFCD00004852
UIJGNTRUPZPVNG-UHFFFAOYSA-N
thiobenzoic acid, benzenecarbothioic acid, benzoyl thiol, monothiobenzoic acid, benzoic acid, thio, acido mercaptobenzoico, unii-gbg5rlo56n, ccris 8913, acido mercaptobenzoico italian, gbg5rlo56n
7414
benzenecarbothioic S-acid
C1=CC=C(C=C1)C(=O)S
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 15.0°C to 18.0°C
Color Yellow to Brown
Density 1.1700g/mL
Boiling Point 122.0°C (30.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Molecular Formula C7H6OS
Linear Formula C6H5COSH
Refractive Index 1.6020 to 1.6080
Quantity 25 g
Beilstein 09, 419
Specific Gravity 1.17
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with organic solvents
Molecular Weight (g/mol) 138.18
Formula Weight 138.18
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Thiobenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • Thiobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.