missing translation for 'onlineSavingsMsg'
Learn More

Thiol-PEG2-acid, TRC

Product Code. 30452839
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. Quantity unitSize
30452839 10 mg 10mg
30453340 5 mg -
30452842 50 mg 50mg
3 options
This item is not returnable. View return policy

Product Code. 30452839

Brand: Toronto Research Chemicals TRCT35122010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1379649-73-6
Molecular Formula C7H14O4S
Molecular Weight (g/mol) 194.25

Specifications

Certifications/Compliance ISO 9001
Shipping Condition Room Temperature
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.