missing translation for 'onlineSavingsMsg'
Learn More

Thiophenol, 99%, AcroSeal™, Thermo Scientific Chemicals

Catalog Number p-4229342
20.46 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
108-98-5
C6H6S
110.17
MFCD00004826
RMVRSNDYEFQCLF-UHFFFAOYSA-N
thiophenol, phenyl mercaptan, mercaptobenzene, phenylthiol, phenylmercaptan, thiofenol, phenol, thio, thiophenate, thiofenol czech, benzene, mercapto
7969
CHEBI:48498
benzenethiol
C1=CC=C(C=C1)S
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
108-98-5
C6H6S
110.17
MFCD00004826
RMVRSNDYEFQCLF-UHFFFAOYSA-N
thiophenol, phenyl mercaptan, mercaptobenzene, phenylthiol, phenylmercaptan, thiofenol, phenol, thio, thiophenate, thiofenol czech, benzene, mercapto
7969
CHEBI:48498
benzenethiol
C1=CC=C(C=C1)S

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -15.0°C
Color Colorless to Yellow
Density 1.0780g/mL
Boiling Point 167.0°C to 169.0°C
Flash Point 50°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging AcroSeal™ Glass bottle
Linear Formula C6H5SH
Refractive Index 1.5880 to 1.5900
Quantity 100 mL
Merck Index 15,9508
Specific Gravity 1.078
Solubility Information Solubility in water: insoluble. Other solubilities: very soluble in alcohol, miscible with ether, benzene and cs2
Viscosity 0.981 mm2/s (30°C)
Formula Weight 110.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Thiophenol
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Thiophenol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 2
  • Acute toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H371-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H361-Suspected of damaging fertility or the unborn child.
  • H372-Causes damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P330-Rinse mouth.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'