missing translation for 'onlineSavingsMsg'
Learn More

THRAP3 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA568012

Additional Details : Weight : 0.01000kg

Product Code. 15873004

  • 472.00€ / 100µL
Estimated Shipment: 27-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Predicted to react with Mouse and Rat samples.

THRAP3 is a 955 amino acid novel nuclear receptor and a member of the Thyroid hormone receptor associated proteins with a LISH domain and no obvious known sequence motifs. THRAP3 is a 150 kDa subunit of the large multiprotein complex TRAP and is important in the maintenance of the fundamental structure of the fibrillar center and dense fibrillar component in the nucleolus. It is also known to play an important role in transcriptional coactivation. It is known to interact with DKC1, nop5/nap65 and fibrillarin. From the Northern blot analysis of several human tissues it was found that TRAP150 is ubiquitously expressed.
Specifications

Specifications

THRAP3
Polyclonal
Unconjugated
THRAP3
9330151F09Rik; B230333E16Rik; BCLAF1 and THRAP3 family member 2; BCLAF2; THRAP3; thyroid hormone receptor associated protein 3; thyroid hormone receptor-associated protein 3; Thyroid hormone receptor-associated protein complex 150 kDa component; thyroid hormone receptor-associated protein, 150 kDa subunit; TRAP150
Rabbit
affinity chromatography
RUO
230753, 313591, 9967
-20°C
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q569Z6, Q5M7V8, Q9Y2W1
THRAP3
Synthesized peptide derived from human THRAP3.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions