Learn More
BOLA Thread Adaptor GL
Product Code. p-200109939
Click to view available options
Thread Size:
GL 14
GL 15
Unit Size:
Each
Product Code. 16224901
Brand: BOLA D53708
Only null left
Add to basket
Add to basket
Thread Adaptor GL PTFE, 2 x Male Thread
  • Straight transition fitting made of PTFE, two GL-threaded connections.
  • Universal chemical resistance, the medium is only exposed to PTFE.
Specifications
Certifications/Compliance FDA
Autoclavable Autoclavable
Thread Size GL 14
Color White