Learn More
BOLA Thread Adaptor GL
Click to view available options
Thread Size:
GL 14
GL 15
Unit Size:
Each
Description
- Straight transition fitting made of PTFE, two GL-threaded connections.
- Universal chemical resistance, the medium is only exposed to PTFE.
Specifications
Specifications
| Certifications/Compliance | FDA |
| Autoclavable | Autoclavable |
| Thread Size | GL 14 |
| Color | White |