Learn More
S-Curve™ Three Compartment Glove Dispenser with Single Lid, Front Access
Click to view available options
Material:
PETG
Description
- Single, sloping lid with full-length hinge to maintain HEPA flow
- Front access holes for easy product dispensing
- Can be mounted with heavy-duty wall brackets (included)
- Flame treated edges for a smooth finish
Specifications
Product Type | Glove Dispenser |
Color | Clear |
Dimensions (D x W x H) Interior | 12 x 16 x 12 in. |
Holds | Gloves |
Material | PETG |
Mounting | Keyhole Slots |
No. of Compartments | 3 |
Product Style | Top Loading |
Includes | Mounting hardware |
Thickness (English) | 0.25 in. |