Learn More
Sampling Systems™ Thumb Tube Pipettes
Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
60 mL
Unit Size:
Pack of 50
Description
- Manufactured with HDPE
- Individually bagged
- FDA conforming materials
- BSE/TSE free
- Full batch traceability
- Available sterilized by gamma
- Press thumb onto the tube until you are at the sampling depth
- Raise the thumb until levels equalise
- Press thumb onto the tube to seal the sample in
Specifications
Specifications
| Material | HDPE |
| Description | Individually Bagged |
| Capacity (Metric) | 60 mL |
| Disposable | Single-use |
| Dimensions (L x W) | 1000 x 12 mm |