missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Thyroglobulin Native Protein

Product Code. 18611728 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18611728 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18611728 Supplier Novus Biologicals™ Supplier No. NBP262539

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Native Protein

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/mL
For Use With (Application) ELISA
Formulation PBS
Gene ID (Entrez) 7038
Molecular Weight (g/mol) MolecularWeight-theroretical: 660 kDa
Quantity 1 mg
Storage Requirements Store at -80°C. Avoid freeze/thaw cycles.
Gene Alias AITD3TGN, TDH3, Tg, thyroglobulin
Gene Symbol TG
Research Category Cancer, Cell Biology
Purity or Quality Grade Affinity purified
Protein Thyroglobulin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.