Learn More
Neogen MonitorMark™ Time Temperature Indicator
Product Code. 17268510
Encompass
Click to view available options
Temperature (Metric) Operating:
10°C
22°C
Unit Size:
Pack of 500
Product Code. 17268510
Brand: Neogen MonitorMark™ 7000001890
This item requires a price update prior to being ordered.
Request a Price for this item here.
Solves the challenge of monitoring thermal exposure of temperature-sensitive products during transportation and storage.
  • Inexpensive solution for monitoring product exposure to temperatures above the specified limit over a 48 hour time period.
  • Self-adhesive backing for easy attachment to secondary packaging.
  • Easily-interpreted visual results.
  • Results indicate both exposure and relative time over which exposure occurred.
  • Removable film activation strip starts the monitoring process on demand for accuracy.
Specifications
Includes 500 /Cs.
Length (English) 3.74 in.
Length (Metric) 95 mm
Response Time 48 Hours
Temperature (English) Operating 50°F
Temperature (Metric) Operating 10°C