missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Timolol Maleate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Timolol Maleate
Specifications
Specifications
| Chemical Name or Material | Timolol Maleate |
| CAS | 26921-17-5 |
| Analyte or Component Names | Timolol Maleate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H24 N4 O3 S . C4 H4 O4 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Timolol Maleate, 2-Propanol, 1-[(1,1-dimethylethyl)amino]-3-[[4-(4-morpholinyl)-1,2,5-thiadiazol-3-yl]oxy]-, (2S)-, (2Z)-2-butenedioate (1:1), 2-Propanol, 1-(tert-butylamino)-3-[(4-morpholino-1,2,5-thiadiazol-3-yl)oxy]-, (-)-, maleate (1:1) (salt) (8CI), 2-Propanol, 1-[(1,1-dimethylethyl)amino]-3-[[4-(4-morpholinyl)-1,2,5-thiadiazol-3-yl]oxy]-, (2S)-, (2Z)-2-butenedioate (1:1) (salt) (9CI), 2-Propanol, 1-[(1,1-dimethylethyl)amino]-3-[[4-(4-morpholinyl)-1,2,5-thiadiazol-3-yl]oxy]-, (S)-, (Z)-2-butenedioate (1:1) (salt), 1,2,5-Thiadiazole, 2-propanol deriv., (-)-Timolol maleate, (S)-(-)-Timolol maleate, (S)-Timolol hydrogen maleate, (S)-Timolol maleate, Aquanil, Betim, Betime, Blocadren, Blocanol, Istalol, L-Timolol maleate, MK 950, Optimol, Proflax, Rysmon TG, Temserin, Tenopt, Timabak, Timacar, Timacor, Timolet, Timolol LA, Timolol hydrogen maleate, Timomann, Timoptic, Timoptol, Timoptol XE, Timorom, WP 934, l-Timolol maleate |
| InChI Formula | InChI=1S/C13H24N4O3S.C4H4O4/c1-13(2,3)14-8-10(18)9-20-12-11(15-21-16-12)17-4-6-19-7-5-17;5-3(6)1-2-4(7)8/h10,14,18H,4-9H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1-/t10-;/m0./s1 |
| SMILES | CC(C)(C)NC[C@H](O)COc1nsnc1N2CCOCC2.OC(=O)\C=C/C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;(2S)-1-(tert-butylamino)-3-[(4-morpholin-4-yl-1,2,5-thiadiazol-3-yl)oxy]propan-2-ol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction