missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tinidazole

Product Code. 16504394 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 16504394

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19387-91-8
Molecular Formula C8H13N3O4S
Molecular Weight (g/mol) 247.27
MDL Number MFCD00057217
InChI Key HJLSLZFTEKNLFI-UHFFFAOYSA-N
IUPAC Name 1-[2-(ethanesulfonyl)ethyl]-2-methyl-5-nitro-1H-imidazole
SMILES CCS(=O)(=O)CCN1C(C)=NC=C1[N+]([O-])=O

Specifications

Melting Point 124.0°C to 128.0°C
Quantity 25 g
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: soluble in acetone, slightly soluble in methanol, very slightly soluble on ethanol
Formula Weight 247.27
Chemical Name or Material Tinidazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.