Learn More
Lovibond™ Tintometer™ Bromocresol Green Tablets
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Brand: Lovibond™ 511760
Additional Details : Weight : 0.30000kg
Description
- Bromocresol Green Tablets
- Available in packs of 100 or 250
Specifications
Photometry Tablet Reagents | |
Test kits, colorimeters and spectrophotometers | |
100 Tablets | |
Bromocresol Green Tablets |