Learn More
Lovibond™ Tintometer™ Bromocresol Green Tablets
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Brand:  Lovibond™ 511761
Additional Details : Weight : 0.30000kg
 View more versions of this product
Product Code. 15391735
  • 76.30€ / Pack of 250
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
Description
Description
  • Bromocresol Green Tablets
  • Available in packs of 100 or 250
Specifications
Specifications
Photometry Tablet Reagents
Test kits, colorimeters and spectrophotometers
250 Tablets
Bromocresol Green Tablets