Learn More
Lovibond™ Tintometer™ Bromophenol Blue Tablets
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Brand:  Lovibond™ 511620
Additional Details : Weight : 0.30000kg
 View more versions of this product
Product Code. 15381725
  • 34.90€ / Pack of 100
Estimated Shipment: 03-06-2024
to see stock.
Add to basket
Description
Description
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
  • Bromophenol Blue Tablets
  • Available in packs of 100 and 250
Specifications
Specifications
Photometry Tablet Reagents
Test kits, colorimeters and spectrophotometers
100 Tablets
Bromophenol Blue Tablets