Learn More
Lovibond™ Tintometer™ CHECKIT™ Comparator, Zinc LR Test, 0 to 1mg/L
Suitable for both mobile and static water analysis. Checkit™ Comparators are available with a number of different color scales and provide the basis for a comprehensive easy to to to use analysis system.
Brand:  Lovibond™ 147340
Additional Details : Weight : 0.46400kg Transport : UN number : 3077 Chem class : 9 Pack group : III
Product Code. 10327052
  • 75.20€ / Each
Estimated Shipment: 17-05-2024
to see stock.
Description
Description
Suitable for both mobile and static water analysis. Checkit™ Comparators are available with a number of different color scales and provide the basis for a comprehensive easy to to to use analysis system.
  • The Lovibond™ Checkit™ comparator is a compact, lightweight colorimetric unit
  • Suitable for both mobile and static water analysis
  • Available with a number of different color scales
  • Provides the basis for a comprehensive easy to to to use analysis system
  • Each Checkit™ Comparator Disc contains a continuous colour scale for exact color matching between standard and sample
  • Each Checkit™ Comparator Kit is supplied in a carry case with comparator, vials, discs, reagents and method instructions for all tests
  • The Zinc disc supplied measures from 0 to 1mg/L with the following steps: 0 / 0.1 / 0.2 / 0.3 / 0.4 / 0.5 / 0.6 / 0.7 / 0.8 / 0.9 / 1.0
Specifications
Specifications
Water Quality Testing
Carry case with Comparator, Vials, Discs, reagents and method instructions for all tests
Zinc LR Test
Checkit™ Comparator
0 to 1mg/L