Learn More
Lovibond™ Tintometer™ DPD 1 and DPD 3 High Calcium Tablets Combi-Pack
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Brand:  Lovibond™ 517782BT
Additional Details : Weight : 0.30000kg
 View more versions of this product
Product Code. 15381975
  • 103.00€ / Pack of 250
Estimated Shipment: 25-04-2024
to see stock.
Add to basket
Description
Description
  • New aluminum foil blister packaging brings added convenience to the tradition of protection achieved in Lovibond’s long established tablet production technology
  • With the new blister strip, the user just pushes the tablet through the protective foil, straight into the sample
  • Each tablet is contained within an individually formed foil cup, lined with the latest aluminum composite material, and guaranteeing product performance
  • Blister pack has been reduced in size to 91 x 34mm for convenient storage and shipping
  • ‘BT’ is added to the end of the code to identify the new style of packaging
  • Simple, time-saving and practical
  • There are no safety risks if the tablets are used in line with the instructions supplied
  • Safety data sheets are available for all reagents
Specifications
Specifications
Photometry Tablet Reagents
Test kits, colorimeters and spectrophotometers
500 Tablets
DPD 1 and DPD 3 High Calcium Tablets Combi-Pack