missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Tintometer™ Phosphate, total LR Tube Test, 0.07 to 3mg/L and 0.2 to 10mg/L

Product Code. 15372045
Click to view available options
Range:
0.07 to 3 mg/L and 0.2 to 10 mg/L
Unit Size:
Each
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 15372045

Brand: Lovibond™ 2419019

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Use these tests with test kits, colorimeters and spectrophotometers

  • Parameter: Total Phosphate
  • Method: Phosphomolybdic acid
  • For use in test kits, colorimeters and spectrophotometers
  • Case of 24
  • Includes:
  • Reaction Tube, LR
  • Phosphate-101
  • 2 Calibration Tubes
  • Phosphate-102 Solution
  • Phosphate-103 Reagent

Specifications

For Use With (Equipment) Test Kits, Colorimeters and Spectrophotometers
Includes Reaction Tube, LR, Phosphate-101, Calibration Tube - 2 ct., Phosphate-102 Solution, Phosphate-103 Reagent
No. of Tests 24
Range 0.07 to 3 mg/L and 0.2 to 10 mg/L
Test Method Phosphomolybdic acid
Type Phosphate, Total LR Tube Test
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • standard solution ammonia
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 3
  • Substances and mixtures corrosive to metals Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H290-May be corrosive to metals.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H317-May cause an allergic skin reaction.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P313-Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : ammonium persulphate, sulphuric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.