Learn More
Sampling Systems™ Tip for PowderThief Sampler 25mm Diameter
Extensively used throughout the pharmaceutical, chemical and food industries around the world. Sampling Systems™ Tip for PowderThief Sampler 25mm Diameter allow a single sampler to be used to accurately sample a range of different volumes.
Brand:  Sampling Systems™ 1030CT-20
Additional Details : Weight : 0.02340kg
 View more versions of this product
Product Code. 15829201
  • 67.30€ / Each
Estimated Shipment: 31-07-2024
to see stock.
Add to basket
Description
Description
  • Made from high quality 316L stainless steel
  • Wide range of sample sizes
  • Wide range of sampler lengths
  • Easy to use
  • A range of tips that let a single sampler be used to accurately sample a range of different volumes
Specifications
Specifications
Tip
20 mL
25 mm
Stainless Steel (316L)
Powder sampling