Learn More
IKA™ Tip XS Box +
Shop All IKA Products
Click to view available options
Volume (Metric):
10 μL
Unit Size:
Pack of 1920
Description
- Includes 4 stackpacks
- Transparent
- With calibration rings
- Biosphere™ plus (sterile, free of DNA, DNase/RNase, PCR inhib., ATP and pyrogens/endotoxins)
Specifications
Specifications
| Volume (Metric) | 10 μL |
| Filtered | Filtered |
| Sterility | Sterile |
| Color | Transparent |
| Features | Calibration Rings |
| Packaging | 96/Box, 4 Boxes/Pk |
| Product Type | Low Retention Filter Tips |