Learn More
Epredia™ Tissue-Loc™ HistoScreen™ Cassettes, Blue
Product Code. 12648346 Shop All Epredia Products
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Pink
Tan
White
Yellow
Unit Size:
Pack of 1000
Pack of 500
Product Code. 12648346
Brand: Epredia™ C1000AQ
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Handle small biopsies and fragile specimens with Epredia™ Tissue-Loc™ HistoScreen™ Cassettes, the ultimate cassettes for specimen security.
  • Saves time, offers the greatest specimen security, and provides ease of use
  • Provides an inner chamber lined with a “see-through" mesh for easy identification of specimens
  • Eliminates the need for messy sponges or lens paper
  • Ensures tissues are safely processed as cassettes will not pop open during processing
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Mesh Pore Insert
Color Aqua
Product Type HistoScreen™ Cassette