Learn More
Thermo Scientific™ Titan3™ Nylon Syringe Filters
Choose these filters for maximum confidence or for high-solids applications.
Brand: Thermo Scientific™ 44526-NC
Additional Details : Weight : 2.50000kg
Description
- Nylon (Polyamide) membrane
- 0.45μm pore size
- For high-solids applications
Specifications
Nylon | |
500 | |
25 mm |
0.45 μm | |
Titan3 |