missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) isopropoxide, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7050184
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
546-68-9
C12H28O4Ti
284.219
MFCD00008871
VXUYXOFXAQZZMF-UHFFFAOYSA-N
titanium tetraisopropanolate, titanium iv isopropoxide, tetraisopropyl orthotitanate, titanium isopropoxide, titanium tetraisopropylate, titanium isopropylate, ti isopropylate, tetraisopropoxytitanium iv, isopropyl orthotitanate, tetraisopropyl titanate
11026
propan-2-olate;titanium(4+)
CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-].[Ti+4]
This item is not returnable. View return policy
546-68-9
C12H28O4Ti
284.219
MFCD00008871
VXUYXOFXAQZZMF-UHFFFAOYSA-N
titanium tetraisopropanolate, titanium iv isopropoxide, tetraisopropyl orthotitanate, titanium isopropoxide, titanium tetraisopropylate, titanium isopropylate, ti isopropylate, tetraisopropoxytitanium iv, isopropyl orthotitanate, tetraisopropyl titanate
11026
propan-2-olate;titanium(4+)
CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-].[Ti+4]

Titanium(IV) isopropoxide is used as a precursor for the preparation of titanium and barium-strontium-titanate thin films. It is useful to make porous titanosilicates and potential ion-exchange materials for cleanup of radioactive wastes. It is an active component of Sharpless epoxidation as well as involved in the synthesis of chiral epoxides. In Kulinkovich reaction, it is involved as a catalyst in the preparation of cyclopropanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 16°C to 20°C
Color Colorless
Density 0.955 g/mL
Boiling Point 232°C
Flash Point 46°C (115°F)
Linear Formula Ti[OCH(CH3)2]4
Refractive Index 1.464
Quantity 5 g
UN Number UN2413
Beilstein 3679474
Sensitivity Moisture sensitive
Merck Index 14,9480
Solubility Information Miscible with anhydrous ethanol,chloroform,ether and benzene.
Formula Weight 284.23
Percent Purity 99.995%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Titanium(IV) isopropoxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'