missing translation for 'onlineSavingsMsg'
Learn More

Titanium, Ti 3-2.5, Grade 9, R56320, 65g, ARMI∣MBH

Product Code. 17978064
Change view
Click to view available options
Quantity:
65 g
Unit Size:
65g
Product Code. Quantity unitSize
17978064 65 g 65g
1 options
This item is not returnable. View return policy

Product Code. 17978064

Brand: ARMI/MBH IARM261EC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ARMI∣MBH produces high-quality metal alloy certified reference materials and set up standards for Spark-OES, ICP, and XRF elemental analysis. The offering includes steel, aluminum, titanium, nickel, cobalt, copper, magnesium, and more.

Titanium, Ti 3-2.5, Grade 9, R56320, 65g
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Boron/Copper/Molybdenum/Tin/Nitrogen/Silicon/Carbon/Chromium/Nickel/Oxygen/Iron/Vanadium/Aluminium/Cobalt/Hydrogen/Manganese/Ruthenium/Tungsten/Yttrium/Palladium/Zirconium/Niobium/Titanium/Phosphorus
CAS 7440-32-6
Quantity 65 g
Form Pieces
Concentration or Composition (by Analyte or Components) Boron 0.0003 %/Copper 0.0025 %/Molybdenum 0.003 %/Tin 0.005 %/Nitrogen 0.006 %/Silicon 0.007 %/Carbon 0.012 %/Chromium 0.016 %/Nickel 0.018 %/Oxygen 0.084 %/Iron 0.18 %/Vanadium 2.51 %/Aluminium 3.05 %/Cobalt (0.0004) %/Hydrogen (0.0005) %/Manganese (0.001) %/Ruthenium (0.001) %/Tungsten (0.001) %/Yttrium (0.001) %/Palladium (0.002) %/Zirconium (0.003) %/Niobium (0.005) %/Titanium (94.1) %/Phosphorus <0.003 %
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.