missing translation for 'onlineSavingsMsg'
Learn More

Titanium, Ti 6-7, R56700, ARMI∣MBH

Product Code. 17909044
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Product Code. Quantity unitSize
17909044 1 Ea. Each
1 options
This item is not returnable. View return policy

Product Code. 17909044

Brand: ARMI/MBH IARM300AG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ARMI∣MBH produces high-quality metal alloy certified reference materials and set up standards for Spark-OES, ICP, and XRF elemental analysis. The offering includes steel, aluminum, titanium, nickel, cobalt, copper, magnesium, and more.

Titanium, Ti 6-7, R56700, Ø35x19mm
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Molybdenum/Manganese/Hydrogen/Zirconium/Nitrogen/Copper/Tantalum/Vanadium/Carbon/Tin/Nickel/Chromium/Silicon/Oxygen/Iron/Aluminium/Niobium/Titanium/Tungsten/Magnesium/Yttrium/Boron/Cobalt/Phosphorus/Palladium
CAS 7440-32-6
Quantity 1 Ea.
Form Disc
Concentration or Composition (by Analyte or Components) Molybdenum 0.0009 %/Manganese 0.0012 %/Hydrogen 0.0018 %/Zirconium 0.003 %/Nitrogen 0.0033 %/Copper 0.0039 %/Tantalum 0.004 %/Vanadium 0.005 %/Carbon 0.007 %/Tin 0.007 %/Nickel 0.0081 %/Chromium 0.0097 %/Silicon 0.018 %/Oxygen 0.162 %/Iron 0.191 %/Aluminium 5.96 %/Niobium 6.86 %/Titanium 86.71 %/Tungsten (<0.002) %/Magnesium (0.0004) %/Yttrium (0.0004) %/Boron (0.0008) %/Cobalt (0.001) %/Phosphorus (0.001) %/Palladium (0.002) %
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.