Learn More
BRAND™ Titrating Tube for Titrette™ Bottletop Burette
Shop All Brand Products
Click to view available options
For Use With (Equipment):
10 mL Titrette
25 mL Titrette
Unit Size:
Each
Description
- For use with Titrette™ Bottle Top Burette
- Screw cap fits onto titrating tube to prevent leakage
- Includes discharge/recirculation valve
Specifications
Specifications
| Product Type | Titrating Tube |
| For Use With (Equipment) | 10 mL Titrette |
| Includes | Screw cap for tube, discharge/recirculation tube |
| Quantity | 1 Ea. |