Learn More
TUTTNAUER Tiva 10 Glassware Washer with Automatic Door
Product Code. 30058121 Shop All Tuttnauer Products
Click to view available options
Voltage:
380/400 V
Unit Size:
1 unit
Regulatory: This product is under eco-contribution tax.
Product Code. 30058121
Brand: TUTTNAUER TIVA101VL
Only null left
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
Equipped with an automatic sliding door, of double tempered glass, for laboratory glassware with 4 independent levels.
  • The upper levels are removable for washing large items.
  • Equipped with a system that filters, electrically heats, and forces air with a powerful blower in the wash chamber and inside hollow instruments for excellent drying results.
  • A wide range of racks, nozzles, and accessories make these machines suitable for washing and drying various laboratory glassware.
Specifications
Display LCD
Material Stainless Steel
Product Type Glassware Washer with Drying System and Steam Condenser
Style Automatic Door
Voltage 380/400 V