missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TL 0259

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
TL 0259 is a selective and highly potent Fgr and Lyn inhibitor (IC50 values are 0.03 and 0.1 nM, respectively). Exhibits selectivity for Fgr and Lyn over other Src family kinases and other nonreceptor and receptor tyrosine kinases. Suppresses proliferation of acute myelogenous leukemia (AML) cell lines in vitro. Eliminates leukemic cells from spleen and bone of mice bearing AML xenografts. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 3-[(6,7-Dimethoxy-4-quinazolinyl)oxy]-N-[4-[(4-ethyl-1-piperazinyl)methyl]-3-(trifluoromethyl)phenyl]-4-methylbenzamide dihydrochloride |
| Quantity | 10 mg |
| Molecular Formula | C32H34F3N5O4.2HCl |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction