missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ TNF-alpha Antibody Pair [HRP]

Product Code. 18215286 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18215286

Brand: Novus Biologicals™ NBP2793905PLATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Ferret TNF-alpha.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 156.25 pg/mL
Assay Sensitivity 156.25-5000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Ferret TNF-alpha.
Target Species Ferret
For Use With (Application) Sandwich ELISA
Gene Alias APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Gene ID (Entrez) 7124
Gene Symbol TNF
Host Species Mouse/Rabbit
Kit Contents
  • Standard: Each vial contains 70 ng of recombinant TNF-alpha. Reconstitute with 1 mL detection antibody dilution buffer.
  • Capture Antibody: 0.5 mg/mL of rabbit anti-TNF-alpha monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL mouse anti-TNF-alpha monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Quantity 5 Plates
    Regulatory Status RUO
    Research Discipline Adaptive Immunity, Asthma, Autophagy, Biologically Active Proteins, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity
    Storage Requirements Storage is content dependent.
    Target TNF-alpha
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: After reconstitution, store at -20 to -80°C in a manual defrost freezer. A six-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 5 ng/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.