missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ TNF (Human) Recombinant Protein

Product Code. 16212320
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µg
50µg
This item is not returnable. View return policy

Product Code. 16212320

Brand: Abnova™ P5879.100ug

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Human TNF (174 a.a.) full-lenth recombinant protein with His tag expressed in Barley grain (Hordeum vulgare). This protein is endotoxins-free and thus optimal to use in stem cell cultures.

Specifications

Gene ID (Entrez) 7124
Name tumor necrosis factor (TNF superfamily, member 2)
Preparation Method Hordeum vulgare (barley) expression system
Quality Control Testing SDS-PAGE Stained with Coomassie Blue
Quantity 100 μg
Storage Requirements Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias DIF, TNF-alpha, TNFA, TNFSF2
Gene Symbol TNF
Cross Reactivity Human
Species Plants
Protein Tag His
Buffer Lyophilized from PBS, pH 7.2
Form Lyophilized
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.