missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tool Steel, AISI S7, T41907, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Tool Steel, AISI S7, T41907, 100g
Specifications
Specifications
| Chemical Name or Material | Boron/Zirconium/Oxygen/Titanium/Niobium/Tin/Arsenic/Nitrogen/Cobalt/Aluminium/Phosphorus/Tungsten/Copper/Nickel/Vanadium/Manganese/Silicon/Carbon/Molybdenum/Chromium/Lead |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Boron 0.0003 %/Zirconium 0.001 %/Oxygen 0.0014 %/Titanium 0.0026 %/Niobium 0.003 %/Tin 0.004 %/Arsenic 0.006 %/Nitrogen 0.0077 %/Cobalt 0.011 %/Aluminium 0.016 %/Phosphorus 0.02 %/Tungsten 0.035 %/Copper 0.081 %/Nickel 0.194 %/Vanadium 0.256 %/Manganese 0.399 %/Silicon 0.44 %/Carbon 0.479 %/Molybdenum 1.43 %/Chromium 3.27 %/Lead <0.0005 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction