Learn More
BINDER™ Top Access Port 80 to 125 mm
Product Code. 15902992 Shop All BINDER Products
Click to view available options
Material:
Steel
Unit Size:
Each
Product Code. 15902992
Brand: BINDER™ 80121533
Only null left
Add to basket
Add to basket
BINDER™ Top Access Port is an accessory for use on the top of a variety of chambers and incubators.
  • For use on top
  • Includes silicone plug
  • For use with a wide range of BINDER™ incubators and chambers
Specifications
Description Access port with silicone plug - 125 mm, top
For Use With (Equipment) BINDER Chambers Series MK
Material Steel
Type Access Port
Includes Silicone plug
Diameter (Metric) Inside 125 mm