missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Top-Line Extended Length Pipette Tips

Product Code. 10468072 Shop All Fisherbrand Products
Click to view available options
Sterility:
Non-sterile
Sterile
Color:
Blue
Clear
Format:
Bulk
Rack
Unit Size:
Pack of 1000
Pack of 960
This item is not returnable. View return policy

Product Code. 10468072

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Eliminate sample contamination and prevent sample loss. . Fisherbrand™ Top-Line Pipette Tips provide precision pipetting with conical tip and secure fit.

  • Optimized cone geometry for a secure fit
  • Graduation marks on tip at standard volumes
  • Conical tip opening prevents drop formation at the end of the tip
  • Tips made of autoclavable, non-wettable, virgin polypropylene
  • Sterile packs available (beta irradiation)

Specifications

Volume (Metric) 100 to 1000 μL
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Length (Metric) 86 mm
Certifications/Compliance ISO Standard 8655, EU-Directive 98/79/EG, competent authority code DE/CA90, EN 552/ISO 11137 and DIN EN 556, EU Directive 2002/95/EC, ISO Standard 14000 and ISO Standard 14001, ISO Standard 10993, DIN Standard 12600
Sterilization Method Autoclave
Format Bulk
Tip Style Extended Length
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line Top-Line
Material Virgin Polypropylene
Graduations Standard volumes
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.