missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Torin 2
GREENER_CHOICE

Product Code. 18015192
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18015192

Brand: Tocris Bioscience™ 4248/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective mTOR inhibitor

Potent and selective mTOR inhibitor (IC50 = 2.1 nM). Displays 800-fold cellular selectivity for mTOR over PI3K (cellular EC50 values are 0.25 and 200 nM for mTOR and PI3K respectively). Activates autophagy. Orally available.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1223001-51-1
Molecular Formula C24H15F3N4O

Specifications

Quantity 10 mg
Formula Weight 432.4
Purity >98%
Molecular Weight (g/mol) 432.406
Chemical Name or Material Torin 2
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.